Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

5-Sulfosalicylic acid dihydrate ≥99.0% (by acidimetry) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 2-Hydroxy-5-sulphobenzoic acid dihydrate2-Hydroxy-5-sulfobenzoic acid dihydrate

M-Clarity™ quality level = MQ200

8.00691.0250 8.00691.1000 8.00691.0100 8.00691.1000 8.00691.0250 8.00691.0100
EM8.00691.1000CS 794 USD InStock
EM8.00691.1000 EM8.00691.0100 EM8.00691.0250 CA8.00691.0100 CA8.00691.0250 CA8.00691.1000
5-Sulfosalicylic acid dihydrate ≥99.0% (by acidimetry) for synthesis, Sigma-Aldrich®
5-Sulphosalicylic acid dihydrate


Formula: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
MW: 254.22 g/mol
Melting Pt: 224 °C
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00149540
CAS Number: 5965-83-3
EINECS: 202-555-6
UN: 2585
ADR: 8,III
Merck Index: 12,09138
https://us-legacy.vwr.com/stibo/web/std.lang.all/90/04/2299004.jpg
Order Now
Specification Test Results
Assay (acidimetric) ≥ 99.0 %
Heavy metals (as Pb) ≤ 0.001 %
Identity (IR) passes test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty