Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethylenetriamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Diethylenetriamine2,2'-Iminodi(ethylamine)DIEN

M-Clarity™ quality level = MQ200

8.03274.1000 8.03274.0100 8.03274.1000
EM8.03274.1000CS 546 USD InStock
EM8.03274.1000 CA8.03274.1000 CA8.03274.0100
Diethylenetriamine for synthesis, Sigma-Aldrich®
Diethylenetriamine


Formula: C₄H₁₃N₃
MW: 103.17 g/mol
MDL Number: MFCD00008171
CAS Number: 111-40-0
UN: 2079
ADR: 8,II
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -35 °C
Molar Mass 103.16 g/mol
Boiling Point 206 - 209 °C (1013 hPa)
Vapor Pressure 0.5 hPa (20 °C)
Flash Point 97 °C
Refractive Index 1.4826 (20 °C, 589 nm)
Explosion Limit 4.4 - 16.1 %(V)
Density 0.95 g/cm3 (20 °C)
pH 12 (100 g/l, H2O, 20 °C)
Ignition Point 325 °C
Synonyms 2,2'-Iminobis (ethylamine), Bis(2-aminoethyl)amine, 2,2'-Diaminodiethylamine
Hill Formula C4H13N3

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty