Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diacetyl for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 2,3-Butanedione

M-Clarity™ quality level = MQ200

8.03528.0100 8.03528.0100 8.03528.0250
EM8.03528.0100CS 3138 USD InStock
EM8.03528.0100 CA8.03528.0100 EM8.03528.0250
Diacetyl for synthesis, Sigma-Aldrich®
Diacetyl


Formula: C₄H₆O₂
MW: 86.09 g/mol
MDL Number: MFCD00008756
CAS Number: 431-03-8
UN: 2346
ADR: 3,II
Order Now
Specification Test Results
Solubility 200 g/l (25 °C)
Melting Point -2 °C
Molar Mass 86.09 g/mol
Boiling Point 89 - 90 °C (1013 hPa)
Vapor Pressure 52 hPa (20 °C)
Flash Point 5 °C
Explosion Limit 2.4 - 13.0 %(V)
Density 0.99 g/cm3 (20 °C)
Ignition Point 365 °C
Synonyms 2,3-Butanedione, 2,3-Butadione, Dimethyldiketon, Dimethylglyoxal
Hill Formula C4H6O2
Chemical Formula CH3COCOCH3

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty