Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Ionone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: (E)-4-(2,6,6-Trimethylcyclohex-2-en-1-yl)but-3-en-2-one

M-Clarity™ quality level = MQ200

8.04778.0050 8.04778.0050
EM8.04778.0050CS 3993 USD InStock
EM8.04778.0050 CA8.04778.0050
α-Ionone for synthesis, Sigma-Aldrich®
α-Ionone


Formula: C₁₃H₂₀O
MW: 192.3 g/mol
MDL Number: MFCD00001565
CAS Number: 127-41-3
Order Now
Specification Test Results
Solubility 110 mg/l (20 °C)
Molar Mass 192.3 g/mol
Boiling Point 237 °C (1013 hPa)
Vapor Pressure 0.0013 hPa (20 °C)
Flash Point 117 °C
Refractive Index 1.5 (20 °C, 589 nm)
Density 0.93 g/cm3 (20 °C)
Hill Formula C13H20O

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty