Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sebaconitrile for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 1,8-DicyanooctaneSebacic acid dinitrile

M-Clarity™ quality level = MQ200

8.14070.0025 8.14070.0025
EM8.14070.0025CS 6802 USD InStock
EM8.14070.0025 CA8.14070.0025
Sebaconitrile for synthesis, Sigma-Aldrich®
Sebaconitrile


Formula: C₁₀H₁₆N₂
MW: 164.25 g/mol
MDL Number: MFCD00019900
CAS Number: 1871-96-1
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 7 - 8 °C
Molar Mass 164.24 g/mol
Boiling Point 199 - 204 °C (20 hPa)
Flash Point 93 °C
Refractive Index 1.4474 (20 °C, 589 nm)
Density 0.91 g/cm3 (20 °C)
Synonyms Decanedinitrile, Decanedioic acid dinitrile, Sebaconitrile
Hill Formula C10H16N2
Chemical Formula CN(CH2)8CN

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty