Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyrazine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

8.07064.0010 8.07064.0100 8.07064.0100
EM8.07064.0100CS 4635 USD InStock
EM8.07064.0100 CA8.07064.0010 CA8.07064.0100
Pyrazine for synthesis, Sigma-Aldrich®
Pyrazine


Formula: C₄H₄N₂
MW: 80.09 g/mol
MDL Number: MFCD00006122
CAS Number: 290-37-9
UN: 1325
ADR: 4.1,II
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 49 - 52 °C
Molar Mass 80.08 g/mol
Boiling Point 115 - 118 °C (1013 hPa)
Synonyms 1,4-Diazine
Hill Formula C4H4N2

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty