Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Piperazinedione for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Piperazine-2,5-dioneGlycine anhydride

M-Clarity™ quality level = MQ200

8.14160.0025 8.14160.0025
EM8.14160.0025CS 11642 USD InStock
CA8.14160.0025 EM8.14160.0025
2,5-Piperazinedione for synthesis, Sigma-Aldrich®
2,5-Piperazinedione


Formula: C₄H₆N₂O₂
MW: 114.1 g/mol
MDL Number: MFCD00006009
CAS Number: 106-57-0
Order Now
Specification Test Results
Solubility 142 g/l (20 °C)
Molar Mass 114.1 g/mol
Synonyms Glycinanhydride, 2,5-Diketopiperazine
Hill Formula C4H6N2O2

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty