Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iminodiacetic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 2,2'-Azanediyldiacetic acid

M-Clarity™ quality level = MQ200

8.14165.0250 8.14165.0250
EM8.14165.0250CS 1912 USD InStock
CA8.14165.0250 EM8.14165.0250
Iminodiacetic acid for synthesis, Sigma-Aldrich®
Iminodiacetic acid


Formula: C₄H₇NO₄
MW: 133.10 g/mol
MDL Number: MFCD00004280
CAS Number: 142-73-4
Order Now
Specification Test Results
Solubility 42 g/l (20 °C)
Melting Point 247 °C (decomposition)
Molar Mass 133.10 g/mol
pH 2.2 - 2.3 (H2O, 20 °C) (saturated solution)
Hill Formula C4H7NO4
Chemical Formula HN(CH2COOH)2

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty