Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(4-Cyanobenzyl)triphenylphosphonium chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ200

8.18511.0005 8.18511.0005
EM8.18511.0005CS 8676 USD InStock
EM8.18511.0005 CA8.18511.0005
(4-Cyanobenzyl)triphenylphosphonium chloride for synthesis, Sigma-Aldrich®
(4-Cyanobenzyl)triphenylphosphonium chloride


Formula: C₂₆H₂₁ClNP
MDL Number: MFCD00011752
CAS Number: 20430-33-5
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 326 - 338 °C
Molar Mass 413.88 g/mol
Synonyms Triphenyl-4-cyanobenzylphosphonium chloride
Hill Formula C26H21ClNP

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty