Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrocinnamaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 2'-Nitrocinnamaldehyde

M-Clarity™ quality level = MQ200

8.18597.0010 8.18597.0010
EM8.18597.0010CS 15238 USD InStock
CA8.18597.0010 EM8.18597.0010
2-Nitrocinnamaldehyde for synthesis, Sigma-Aldrich®
2-Nitrocinnamaldehyde


Formula: C₉H₇NO₃
MDL Number: MFCD00007188
CAS Number: 1466-88-2
Order Now
Specification Test Results
Solubility (20 °C) slightly soluble
Melting Point 126 - 129 °C
Molar Mass 177.15 g/mol
Hill Formula C9H7NO3
Chemical Formula 2-(NO2)C6H4CH=CHCHO

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty