Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzoin ethyl ether for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Benzoin Ethyl EtherO-Ethylbenzoin

M-Clarity™ quality level = MQ200

8.20022.0050 8.20022.0050
EM8.20022.0050CS 1512 USD InStock
EM8.20022.0050 CA8.20022.0050
Benzoin ethyl ether for synthesis, Sigma-Aldrich®
Benzoin ethyl ether


Formula: C₁₆H₁₆O₂
MW: 240.3 g/mol
MDL Number: MFCD00009242
CAS Number: 574-09-4
Order Now
Specification Test Results
Solubility (20 °C) slightly soluble
Melting Point 56 - 59 °C
Molar Mass 240.30 g/mol
Synonyms Benzoin ethyl ether, 2-Ethoxy-2-phenylacetophenone
Hill Formula C16H16O2
Chemical Formula C6H5CHO(C2H5)COC6H5

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty