Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromononane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 1-Nonyl bromide

M-Clarity™ quality level = MQ200

8.21710.0100 8.21710.0100
EM8.21710.0100CS 1326 USD InStock
CA8.21710.0100 EM8.21710.0100
1-Bromononane for synthesis, Sigma-Aldrich®
1-Bromononane


Formula: BrH₂C(CH₂)₇CH₃
MW: 207.15 g/mol
MDL Number: MFCD00000278
CAS Number: 693-58-3
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Molar Mass 207.15 g/mol
Boiling Point 201 °C (1013 hPa)
Flash Point 90 °C
Refractive Index 1.454 (20 °C, 589 nm)
Density 1.09 g/cm3 (20 °C)
Synonyms Nonyl bromide
Hill Formula C9H19Br
Chemical Formula CH3(CH2)8Br

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty