Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(3-Aminopropyl)triethoxysilane, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 3-(Triethoxysilyl)propylamineGeniosil GF 933-AminopropyltriethoxysilaneTriethoxy(3-aminopropyl)silane

M-Clarity™ quality level = MQ200

8.21619.0500 8.21619.0500
EM8.21619.0500CS 1764 USD InStock
CA8.21619.0500 EM8.21619.0500
(3-Aminopropyl)triethoxysilane, Sigma-Aldrich®
(3-Aminopropyl)triethoxysilane


Formula: H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
MW: 221.37 g/mol
MDL Number: MFCD00008207
CAS Number: 919-30-2
Order Now
Specification Test Results
Molar Mass 221.37 g/mol
Synonyms 3-Aminopropyltriethoxysilane
Chemical Formula (C2H5O)3Si(CH2)3NH2

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty