Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methylamine 40% in water for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger

M-Clarity™ quality level = MQ200

8.22091.2500 8.22091.2500 8.22091.1000 8.22091.1000
EM8.22091.1000CS 276 USD InStock
EM8.22091.1000 EM8.22091.2500 CA8.22091.1000 CA8.22091.2500
Methylamine 40% in water for synthesis, Sigma-Aldrich®
Methylamine


Formula: CH₅N
MW: 31.06 g/mol
MDL Number: MFCD00008104
CAS Number: 74-89-5
UN: 1235
ADR: 3 (8),II
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -40 °C
Boiling Point 48 °C
Vapor Pressure 370 hPa (25 °C)
Flash Point -18 °C
Explosion Limit 5.2 - 26.4 %(V)
Density 0.90 g/cm3 (20 °C)
pH 14 (H2O, 20 °C)
Ignition Point 425 °C

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty