Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9-Vinylanthracene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ200

8.24593.0500 8.24593.0500
EM8.24593.0500CS 153822 USD InStock
CA8.24593.0500 EM8.24593.0500
9-Vinylanthracene for synthesis, Sigma-Aldrich®
9-Vinylanthracene


Formula: C₁₆H₁₂
MW: 204.27 g/mol
MDL Number: MFCD00003576
CAS Number: 2444-68-0
Order Now
Specification Test Results
Melting Point 61 - 64 °C
Molar Mass 204.27 g/mol
Synonyms 9-Ethenylanthracene
Hill Formula C16H12

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty