Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vanillic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 4-Hydroxy-3-methoxybenzoic acid

M-Clarity™ quality level = MQ200

8.41025.0050 8.41025.0050
EM8.41025.0050CS 2791 USD InStock
EM8.41025.0050 CA8.41025.0050
Vanillic acid for synthesis, Sigma-Aldrich®
Vanillic acid


Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00002551
CAS Number: 121-34-6
Order Now
Specification Test Results
Solubility (20 °C) slightly soluble
Melting Point 208 - 210 °C
Molar Mass 168.14 g/mol
Synonyms 4-Hydroxy-3-methoxybenzoic acid
Hill Formula C8H8O4

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty