Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Methylglutaric acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 2-Methylpentanedioic acid

M-Clarity™ quality level = MQ200

8.41328.0005 8.41328.0005
EM8.41328.0005CS 62363 USD InStock
CA8.41328.0005 EM8.41328.0005
(±)-2-Methylglutaric acid for synthesis, Sigma-Aldrich®
(±)-2-Methylglutaric acid


Formula: C₆H₁₀O₄
MW: 146.14 g/mol
MDL Number: MFCD00002661
CAS Number: 617-62-9
Order Now
Specification Test Results
Melting Point 74 - 76 °C
Molar Mass 146.14 g/mol
Synonyms 2-Methylpentanedioic acid
Hill Formula C6H10O4

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty