Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylfluorone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 9-Phenyl-3-fluorone9-Phenyl-2,3,7-trihydroxy-6-fluorone2,6,7-Trihydroxy-9-phenyl-3-isoxanthone

M-Clarity™ quality level = MQ200

8.41569.0001 8.41569.0001 8.41569.0005 8.41569.0005
EM8.41569.0001CS 25342 USD InStock
CA8.41569.0001 CA8.41569.0005 EM8.41569.0001 EM8.41569.0005
Phenylfluorone for synthesis, Sigma-Aldrich®
Phenylfluorone


Formula: C₁₉H₁₂O₅
MW: 320.30 g/mol
MDL Number: MFCD00005048
CAS Number: 975-17-7
Order Now
Specification Test Results
Solubility 300 °C
Melting Point 320.3 g/mol
Hill Formula C19H12O5

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty