Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-C6orf70 Rabbit Polyclonal Antibody

Supplier: Proteintech
24690-1-AP-150UL
10155-278EA 453 USD InStock
10155-278
Anti-C6orf70 Rabbit Polyclonal Antibody
Antibodies
The C6orf70 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human C6orf70. This antibody recognizes human antigen. The C6orf70 antibody has been validated for the following applications: ELISA, IHC analysis.

WB 1 : 300 ; IHC 1 : 50 ; IF 1 : 25

Type: Primary
Antigen: C6orf70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty