Would you like to log in again?
BioFX® 450 nm Stop Reagent for TMB Microwell Substrates, Surmodics
Supplier: Surmodics
BioFX®
STPR-0100-01 STPR-1000-01
100359-302EA 150 USD InStock
100359-302 100359-300
BioFX® 450 nm Stop Reagent for TMB Microwell Substrates, Surmodics
Assays ELISAs
Absorbance is read in the yellow spectral range (450 nm).
  • 3 year shelf life at room temp
A dry blend stop reagent used in end point ELISA applications where a TMB substrate is used as the detection mechanism.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty