Would you like to log in again?
BioFX® Liquid Stop Solution for TMB Microwell Substrates, 450 nm, Surmodics
Supplier: Surmodics
BioFX®
LSTP-1000-01 LSTP-0100-01
100359-306EA 140 USD InStock
100359-306 100359-304
BioFX® Liquid Stop Solution for TMB Microwell Substrates, 450 nm, Surmodics
Assays ELISAs
A liquid stop solution used in end point ELISA applications where a TMB substrate is used as the detection mechanism.
  • Ready to use
  • 3 year shelf life
Absorbance is read in the yellow spectral range (450 nm).
Certifications: ISO 13485:2016 & 9001:2015 Certified
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty