Would you like to log in again?
Proteinase K
Supplier: New England Biolabs
P8107S P8107S
101076-584EA 96 USD InStock
101076-584 CA101076-584
Proteinase K
Enzymes
Highly characterized for more consistent performance, Proteinase K is a subtilisin-related serine protease that will hydrolyze a variety of peptide bonds
Proteinase K is active in a wide range of temperatures and buffers with optimal activity between 20 and 60°C and a pH between 7.5 and 12.0. Activity is stimulated when up to 2% SDS or up to 4 M urea are included in the reaction. Calcium is important for thermostability of Proteinase K but it is not required for catalysis, therefore Proteinase K is also active in buffers containing chelating agents such as EDTA.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty