Would you like to log in again?
Recombinant PNGase F (Glycerol-Free)
Supplier: New England Biolabs
P0709S P0709S P0709L P0709L
102877-614EA 184 USD InStock
102877-614 CA102877-606 102877-606 CA102877-614
Recombinant PNGase F (Glycerol-Free)
Enzymes
Peptide-N-Glycosidase F, also known as PNGase F, is a recombinant amidase which is supplied glycerol free for optimal performance in HPLC intensive methods
PNGase F cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides from N-linked glycoproteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty