Would you like to log in again?
beta-1-4 Galactosidase S
Supplier: New England Biolabs
P0745S P0745L P0745S P0745L
102877-628EA 609 USD InStock
CA102877-628 102877-628 CA102877-630 102877-630
beta-1-4 Galactosidase S
Enzymes
β1-4 Galactosidase S is a highly specific exoglycosidase that catalyzes the hydrolysis of β1-4 linked galactose residues from oligosaccharides.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty