Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lactophenol blue solution stain (fungal), Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Lactophenol anilin blue solutionLactophenol cotton blue solution

M-Clarity™ quality level = MQ300

1.13741.0100
EM1.13741.0100EA 68 USD InStock
EM1.13741.0100
Lactophenol blue solution stain (fungal), Sigma-Aldrich®
Lactophenol blue solution
Lactophenol blue solution for staining fungi.

Lactophenol blue solution is ready-to-use staining solution for fungi staining in bacteriology with samples of human origin. The material is stained in one step with Lactophenol blue solution. Fungi appear dark blue. This ready-to-use solution is especially for air-dried, heat-fixed smears of bacteriological material, e.g. sputum, FNAB, lavages, imprints, body fluids, puss, and exudates, liquid and solid cultures.
The staining solution is sufficient for up to 50 preparations per bottle. It is developed and registered as CE / IVD product for applications in diagnostics.


Density: 1.16 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00146574
UN: 2821
ADR: 6.1,
Order Now
Specification Test Results
Density 1.16 g/cm³ (20 °C)
pH value 2.3 (H₂O, 20 °C)
Solubility (20 °C) soluble
Storage class 6.1B Non combustible substances, toxic
WGK WGK 2 water endangering
R Phrase R 23/24/25-34-48/20/21/22-68
S Phrase S 26-36/37/39-45
Storage Store at +15°C to +25°C.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty