Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Butanesulfonic acid sodium salt, Supelco®

Supplier: MilliporeSigma
Synonyms: Butane-1-sulphonic acid sodium saltButane-1-sulfonic acid sodium saltSodium 1-butanesulphonateSodium 1-butansulfonateSodium 1-butansulphonateSodium 1-butanesulfonateSodium butane-1-sulfonateSodium butane-1-sulphonate

M-Clarity™ quality level = MQ100

1.18303.0025 1.18303.0025
EM1.18303.0025CS 9317.05 USD InStock
CA1.18303.0025 EM1.18303.0025
1-Butanesulfonic acid sodium salt, Supelco®
1-Butansulphonic acid sodium salt
Butane-1-sulfonic acid sodium salt for ion pair chromatography LiChropur®.


Formula: C₄H₉NaO₃S
MW: 160.17 g/mol
MDL Number: MFCD00007540
CAS Number: 2386-54-1
Order Now
Specification Test Results
Hill Formula C₄H₉NaO₃S
Molar Mass 160.17 g/mol
Melting Point 310 °C
Storage class 10 - 13 Other liquids and solids
WGK WGK 3 highly water endangering

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty