Would you like to log in again?
AlphaTec® 88-392 Natural Rubber Latex Gloves, Ansell
Supplier: Ansell
AlphaTec®
193926 113977 113975 113973 193928 113970 113971 193922 113974
32890-367CS 419.43 USD InStock
32890-367 32890-364 76169-889 32890-363 32890-362 32890-361 32890-360 76169-887 32890-357
AlphaTec® 88-392 Natural Rubber Latex Gloves, Ansell
Gloves
An ideal glove for safety and dexterity in the food processing industry, these gloves provide excellent dexterity, tensile strength and elasticity.
  • Fish scale pattern: Excellent grip on wet and dry surfaces
  • Superior dexterity and tensile strength
  • Special chlorination treatment, lessening the threat of related allergy risks
The 20-millimeter thickness provides good chemical, snag, puncture and abrasion resistance.
Use in the agriculture, viticulture, food processing, and food service industries, as well as in general industry.
Uses include canning and food processing; food packing and handling; fruit and vegetable processing; light-duty maintenance; and clean up.
Certifications: Complies with FDA food handling requirements (21 CFR 177.2600).
Packaging: Gloves are packaged 12 pairs in a bag; 144 in a carton.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty