Would you like to log in again?
1000 A Clamp Meter with IR Thermometer, Extech
Supplier: Extech
CM78 CM78-NIST
10748-280EA 1429.41 USD InStock
10748-280 10748-278
1000 A Clamp Meter with IR Thermometer, Extech
Electrical Meters Voltmeters
The FLIR CM78 is a True RMS Industrial Grade 1000A Clamp Meter for the electrician who needs a safe, capable combination tool engineered with advanced power analysis and variable frequency drive filtering functions required by electrical troubleshooters
  • IR thermometer included
  • Bluetooth capable
Clamp Meter for the electrician who needs a safe, capable combination tool.
Certifications: UL
Ordering information: Complete with CAT IV Silicone Test Leads, 6 AAA Batteries, User Manual/CD, and Limited Lifetime Warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty