Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bacon Yummies™ Treats, Certified, Bio-Serv

Supplier: Bio-Serv
Bacon Yummies™
F05778 F05778-1 S05778-1 F05778 S05778-1 F05778-1
89067-258EA 193.72 USD InStock
CA89067-256 89067-258 CA89067-258 89067-256
Bacon Yummies™ Treats, Certified, Bio-Serv
Animal Supplies Animal Feed
Nutritionally complete 190 mg grain-based treats for rodents and supplements for canines and ferrets are excellent for reward or foraging.

  • Can be fed as a sole source diet for rodents with poor appetites
  • Highly palatable bacon flavor
  • Nutritionally assayed
  • Shelf Life: 6 months, cool dry storage

Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Bacon Flavor, Magnesium Stearate, Calcium Silicate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin.

Caloric value: 3.37 kcal/g
Nutritional ash content: 7.8%
Nutritional carbohydrate content: 53.5%
Nutritional fat content: 4.3%
Nutritional fibre content: 4.0%
Nutritional moisture content: <10%
Nutritional protein content: 21.3%

Certifications: Certified (Contaminant Screened).

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty