Would you like to log in again?
Nalgene® Syringe Prefilter, Glass Fiber, 25mm, Thermo Scientific
Supplier: Thermo Scientific
722-2000
10218-466CS 211.7 USD InStock
10218-466
Nalgene® Syringe Prefilter, Glass Fiber, 25mm, Thermo Scientific
Filters Syringe Filters
Clarify highly viscous solutions and remove large particulates from suspensions using this syringe prefilter.
  • Nonsterile
  • Autoclavable
  • Clear housing ring
Can increase overall filtration performance by up to 50% when used in serial filtration.Glass fiber prefilter in polypropylene housing with female Luer-Lok® inlet and male Luer-slip outlet.
Packaging: Bulk packed in zip closure bag.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty