Would you like to log in again?
Drum Labels, Brady
Supplier: Brady
60364 60364
12000-886PK 120.93 USD InStock
12000-886 CA12000-886
Drum Labels, Brady
Labels
These durable labels are used to easily identify and sort waste for disposal.
  • Labels remove cleanly, leaving no residue on surface
Made of coated Paper with pressure sensitive acrylic adhesive which can adheres to most surfaces.
Certifications: Meet D.O.T. requirements HM-181 regulation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty