Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse Diet, Helicobacter 4 Drug Combo, Sterile, Bio-Serv

Supplier: Bio-Serv
S4998 S4998
89408-222BX 519.7 USD InStock
89408-222 CA11008-342
Mouse Diet, Helicobacter 4 Drug Combo, Sterile, Bio-Serv
Animal Supplies Animal Feed
The most effective oral treatment against a broad spectrum of Helicobacter strains

  • Highly palatable, bacon-flavored, nutritionally complete diet
  • Used to treat breeders or quarantined mice
  • Alternative method of treatment to cesarean rederivation
  • Comprehensive husbandry plan must be employed for success; contact Bio-Serv for details
  • Nutritionally assayed
  • Shelf Life: 6 months, cool dry storage

Ingredients: Ground Corn, Dehulled Soybean Meal, Dried Beet Pulp, Fish Meal, Ground Oats, Brewers Dried Yeast, Cane Molasses, Dehydrated Alfalfa Meal, Dried Whey, Wheat Germ, Porcine Animal Fat (preserved with BHA), Porcine Meat Meal, Wheat Middlings, Mineral Mix, Vitamin Mix, Bacon Flavor, DL-Methionine, Choline Chloride, Omeprazole, Metronidazole, Amoxicillin, Clarithromycin.

5 g of Mouse Helicobacter Diet contains 3.00 mg of Amoxicillin, 0.50 mg of Clarithromycin, 1.00 mg of Metronidazole, and 0.02 mg of Omeprazole. Feed ab lib.

Caloric value: 3.37 kcal/g
Nutritional ash content: 6.9%
Nutritional carbohydrate content: 48.4%
Nutritional fat content: 5.4%
Nutritional fibre content: 5.1%
Nutritional moisture content: <10%
Nutritional protein content: 23.7%

Ordering information: Minimum Order is 10 kg; sold in 10 kg increments.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty