Would you like to log in again?
Coated Paper GHS Labels 'Corrosive Picto', Brady
Supplier: Brady
118895 118894 118893
53519-906PK 15.26 USD InStock
53519-906 53519-904 53519-902
Coated Paper GHS Labels 'Corrosive Picto', Brady
Labels
Brady B-235 coated paper with pressure sensitive acrylic adhesive adheres to most surfaces.
  • Permanent adhesive and acrylic
  • Abrasion resistant
  • Chemical resistant
  • Semi-gloss finish
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty