Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Love Mash™ Rodent Reproductive Diet, Sterile, Bio-Serv

Supplier: Bio-Serv
Love Mash™
S3823P S3823P
89067-434EA 50.36 USD InStock
89067-434 CA89067-434
Love Mash™ Rodent Reproductive Diet, Sterile, Bio-Serv
Animal Supplies Animal Feed
Love Mash is a supplemental diet specifically designed to support reproductive performance in rats and mice.

  • Love Mash should be fed as a supplement to animal's standard diet
  • About two pellets every other day placed at cage floor level recommended
  • Rodents look forward to the treat, and it provides an opportunity for foraging, which decreases stress
  • Start to feed the Love Mash two weeks prior to breeding and continue through weaning

Ingredients: Oatmeal, Cod Liver Oil, Wheat Germ, Brewer's Yeast, Corn Syrup.

Feeding Love Mash increases litter size, improves fertility in both males and females and increases pup survival by decreasing mom's stress and increasing milk production. It has been successful in the most difficult breeders, including transgenics and knockouts. The product's success is a result of the simple formulation, which relies on the fat composition of the diet paired with method of delivery (foraging). It has been successfully used at many large breeding facilities. Nutritionally assayed. Soft pellets of random lengths, 1/2" diameter. Shelf Life: 6 months, refrigerated.

Caloric value: 3.83 kcal/g
Nutritional ash content: 3.0%
Nutritional carbohydrate content: 52.9%
Nutritional fat content: 14.5%
Nutritional fibre content: 9.0%
Nutritional moisture content: <10%
Nutritional protein content: 13.6%

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty