Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rodent Diet, Grain-Based, Fenbendazole 150 mg/kg, Bio-Serv

Supplier: Bio-Serv
S3867 S3867
89408-274EA 299.05 USD InStock
CA89408-274 89408-274
Rodent Diet, Grain-Based, Fenbendazole 150 mg/kg, Bio-Serv
Animal Supplies Animal Feed
A proven effective and safe treatment for pinworms

  • Nutritionally complete, grain-based rodent diet containing 150 mg of Fenbendazole per kg per diet
  • Sterilized by gamma irradiation
  • Nutritionally assayed
  • Shelf Life: 6 months, cool dry storage

Ingredients: Ground Corn, Dehulled Soybean Meal, Dried Beet Pulp, Fish Meal, Ground Oats, Brewers Dried Yeast, Cane Molasses, Dehydrated Alfalfa Meal, Dried Whey, Wheat Germ, Porcine Animal Fat (preserved with BHA), Porcine Meat Meal, Wheat Middlings, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride, Fenbendazole.

Caloric value: 3.35 kcal/g
Nutritional ash content: 7%
Nutritional carbohydrate content: 48.7%
Nutritional fat content: 5%
Nutritional fibre content: 5.1%
Nutritional moisture content: <10%
Nutritional protein content: 23.9%

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty