Would you like to log in again?
Mouse Arch™, Certified, Bio-Serv
Supplier: Bio-Serv
Mouse Arch™
K3454 K3357 K3357 K3454 K3455 K3455
89412-066EA 4.21 USD InStock
89412-066 CA76353-812 76353-756 CA76353-756 CA89412-066 76353-812
Mouse Arch™, Certified, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Compact rodent sheltering device made of high-temp polycarbonate
Excellent quality, yet highly economical. Easy to clean, store and stack. Enhances both breeding and enrichment programs by providing privacy, security and warmth. Autoclavable. Cagewasher safe.
Certifications: Certified (Contaminant Screened).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty