Would you like to log in again?
Quick Link, Stainless Steel, Bio-Serv
Supplier: Bio-Serv
9133 9133
89412-094EA 3.81 USD InStock
89412-094 CA89412-094
Quick Link, Stainless Steel, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Quick Link is made of stainless steel
Use to suspend various enrichment devices. Autoclavable. Cagewasher safe.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty