Would you like to log in again?
Rest Hop™ Rodent Environmental Enrichment, Certified, Bio-Serv
Supplier: Bio-Serv
Rest Hop™
K3550 K3550
89412-102EA 46.12 USD InStock
89412-102 CA89412-102
Rest Hop™ Rodent Environmental Enrichment, Certified, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made of high-temp polycarbonate, Rest Hop is designed to fulfill the 2010 Guide recommendations for greater mobility
The dual purpose shelter allows animals to hide or rest on top. Autoclavable. Cagewasher safe.
Certifications: Certified (Contaminant Screened).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty