Would you like to log in again?
Precious Gem™ Enrichment Device, Certified, Bio-Serv
Supplier: Bio-Serv
Precious Gem™
K3650 K3650
89412-124EA 10.31 USD InStock
89412-124 CA89412-124
Precious Gem™ Enrichment Device, Certified, Bio-Serv
Animal Supplies Animal Environmental Enrichment
The Precious Gem is made of polyurethane, which makes it simple to clean and more durable than rubber
The enrichment device is suitable for multiple species. Dogs enjoy the texture, rabbits are intrigued by the color and primates enjoy foraging for treats placed in the center. This is a toy, which allows for versatility in its use to prevent boredom in the animals. Freeze it in PRANG™ or slather it with peanut butter, and sprinkle seeds, dried fruit, ground food, or foraging crumbles inside. The Precious Gem is a versatile product, as it promotes several natural animal behaviors including foraging, playing, gnawing and exploration, which are essential to good animal health. Cagewasher safe. Manufactured exclusively by Bio-Serv.
Certifications: Certified (Contaminant Screened).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty