Would you like to log in again?
Corning® Gosselin™ Stackable Petri Dishes, 100×15 mm, Corning
Supplier: Corning
Corning®
BP94S-01
10416-320CS 355.47 USD InStock
10416-320
Corning® Gosselin™ Stackable Petri Dishes, 100×15 mm, Corning
Culture Dishes Petri Dishes
The new Corning® Gosselin™ 100x15 Petri dishes design had been designed in collaboration with manual end users.
  • Manufactured from optically clear virgin polystyrene
  • Sterilized by irradiation SAL 10-3
  • Supplied with 4 vents to provide consistent gas exchange
  • New design to enhance stacked dishes stability
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty