Would you like to log in again?
Silver Talon® Cut Resistant Gloves, Wells Lamont
Supplier: Wells Lamont
Whizard®
134527 134528
32935-164EA 28.5 USD InStock
32935-164 32935-162
Silver Talon® Cut Resistant Gloves, Wells Lamont
Gloves
These gloves made of high performance fibers and stainless steel which provides protection against cut and slashes.
  • Unmatched combination of cut resistance, dexterity and durability
  • Unique colored cuff is highly visible
  • Ambidextrous
  • Gauge 10
The patented yarn composition combines high strength, high performance materials with stainless steel, creating excellent tensile strength and superior cut protection. The Silver Talon is also available with a leather palm or urethane grip pattern for applications requiring precise gripping capabilities.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty