Would you like to log in again?
Gold Test, Spectroquant®, MilliporeSigma
Supplier: MilliporeSigma
Spectroquant®
1.14821.0002 1.14821.0002
EM1.14821.0002EA 1696 USD InStock
EM1.14821.0002 CA1.14821.0002
Gold Test, Spectroquant®, MilliporeSigma
Environmental Test Kits
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Highly stable, ready to use reagent mixtures in a user friendly design
  • Free selection of appropriate cell format according to the measuring range
  • Long shelf life up to 3 years - all at room temperature
  • Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty