Would you like to log in again?
Shipper, Category B Frozen, 4 Large Canisters, Therapak®
Supplier: Therapak
37809
10029-274CS 466.47 USD InStock
10029-274
Shipper, Category B Frozen, 4 Large Canisters, Therapak®
Mailers and Shippers
Frozen specimen shipping system includes: transport box (OD: 18x16x14"), large cooler (ID: 16x13x11"), 4 large 95kPa canisters, 12 Aqui-Pak™ segmented absorbent pouches, cushioning material, list of contents card, dry ice label, and required labels for IATA PI650 marking and compliance
Holds up to seventy-two tubes for overnight shipments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty