Would you like to log in again?
ProKlenz™ Booster Sterile Detergent, STERIS®
Supplier: Steris
ProKlenz®
1S1337WR 1S1308WR 1S1308WR 1S1337WR
10755-304CS 1000 USD InStock
CA75780-052 CA75780-060 10755-304 10755-306
ProKlenz™ Booster Sterile Detergent, STERIS®
Cleaning Agents Detergents and Surface Cleansers
Concentrated detergent for controlled environments.
ProKlenz™ Booster Sterile Detergent is formulated with hydrogen peroxide and is designed to remove organic and inorganic residues from product and non-product contact surfaces.
Sterile filtered and filled in a ISO Class 5 aseptic environment, each package is double bagged for easy entry into controlled environments and aseptic core areas. These products have an extensive documentation package to meet validation objectives and are supported by the STERIS® Technical Services team for individualized application and validation assistance.
ProKlenz™ Booster Sterile Detergent will remove residue from product contact surfaces, room surfaces, equipment, gloves, etc. Detergent is supplied in 2.6oz. or 1gal. bottle.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty