Would you like to log in again?
Angiotensin II ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-204
10049-228EA 885.68 USD InStock
10049-228
Angiotensin II ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive ELISA, detecting as little as 4.6 pg/ml
For the quantitative determination of species independent Angiotensin II in plasma and serum samples.
The Angiotensin II EIA kit is a colorimetric, competitive immunoassay kit with results in 3.5 hours.
Ordering information: Contains Microtiter Plate, Conjugate, Antibody, Assay Buffer 16, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, Streptavidin-HRP.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty