Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
LVV Hemorphin 7 ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-205
10049-230EA 550.92 USD InStock
10049-230
LVV Hemorphin 7 ELISA Kit, Enzo Life Sciences
Assays ELISAs
Most sensitive and rapid LVV Hemorphin 7 kit for neuroscience market.
For the quantitative determination of species independent LVV Hemorphin 7 in serum and tissue homogenate samples.
The LVV Hemorphin 7 EIA kit is a colorimetric, competitive immunoassay kit with results in just 3 hours.
Ordering information: Contains Microtiter Plate, Conjugate, Antibody, Assay Buffer 16, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, Streptavidin-HRP.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty