Would you like to log in again?
24(S)-Hydroxycholesterol ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
900-210-0001
10049-240EA 738.22 USD InStock
10049-240
24(S)-Hydroxycholesterol ELISA Kit, Enzo Life Sciences
Assays ELISAs
Only quantitative 24(S)-hydroxycholesterol ELISA kit available for neurodegenerative research.
For the quantitative determination of species independent 24(S)-Hydroxycholesterol in serum, plasma, cerebral spinal fluid, culture supernatants and tissue samples.
The 24(S)-Hydroxycholesterol ELISA kit is a complete, colorimetric, competitive immunoassay kit for the quantitative determination of 24(S)-Hydroxycholesterol in serum, plasma, tissue culture media (culture supernatant), cerebral spinal fluid and tissue homogenate samples with results in just 2 hours.
Ordering information: Contains Microtiter Plate, Conjugate, Antibody, Assay Buffer, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, Streptavidin-HRP.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty