Would you like to log in again?
Shipper, Category B Refrigerated, 2...8 °C, Therapak®
Supplier: Therapak
38028
10029-326CS 401.91 USD InStock
10029-326
Shipper, Category B Refrigerated, 2...8 °C, Therapak®
Mailers and Shippers
Refrigerated Shipping System has been field tested for 2 - 8° C temperature maintenance for 48 hours
System includes: 12 x 12 x 12” 200# corrugated outer box, EPS cooler with 21/2” all around wall thickness, ambient gel wrap, 95kPa specimen transport bag with absorbent to hold up to six specimen tubes, refrigerant packs, list of contents card, filler material, and required labels for IATA PI650 marking and compliance. Packing Instructions are included to facilitate use for safe and compliant shipping.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty