Would you like to log in again?
TearTech® Shipper, EHS Ambient, Therapak®
Supplier: Therapak
56871
10029-392CS 756.42 USD InStock
10029-392
TearTech® Shipper, EHS Ambient, Therapak®
Mailers and Shippers
The TearTech® EHS specimen shipper features a bulk-pack case of shipping supplies to transport ambient samples.
The shipper includes 50 easy to assemble TearTech® boxes, 100 zip-style biohazard transport bags (two pocket bag for specimen sample and requisition form), 100 Aqui-Pak™ segmented absorbent pouches (each Aqui-Pak™ holds up to 6 standard blood or urine tubes), 50 biohazard labels, 50 Exempt Human Specimen labels, and cushioning material where applicable.
The system features the TearTech® EHS shipper box that holds up to four specimen bags consisting of patient samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty