Would you like to log in again?
ADMA (Human) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-323-KI01
10049-638EA 1713.53 USD InStock
10049-638
ADMA (Human) ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of asymmetrical dimethylarginine (ADMA) in human serum, citrate- and EDTA-plasma
Does not cross-react with SDMA, NMMA or L-arginine.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty